Please login to your online account to display your discounted pricing

Alfa Aesar™ 2',4'-Difluoropropiophenone, 98%

$76.50 - $1,028.24

Chemical Identifiers

Chemical Name or Material 2',4'-Difluoropropiophenone
CAS 85068-30-0
Molecular Formula C9H8F2O
Formula Weight 170.16
MDL Number MFCD00015507
Beilstein 7633370
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1203206

 
alfa aesar™
A1203206
5g Each for $76.50

AAA1203214

 
alfa aesar™
A1203214
25g Each for $315.55

AAA1203222

 
alfa aesar™
A1203222
100g Each for $1,028.24
Description & Specifications

Specifications

Formula Weight 170.16
Chemical Name or Material 2',4'-Difluoropropiophenone
Assay 98%
Molecular Formula C9H8F2O
MDL Number MFCD00015507
CAS 85068-30-0
Beilstein 7633370
Flash Point 76°C (168°F)
Boiling Point 51° to 52°C (1.5mmHg)
Quantity 5g