Learn More
2,4-Difluorophenylboronic acid, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 359130500
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
2, 4-Difluorophenylboronic acid | |
97.5 | |
Authentic | |
Glass bottle | |
50g | |
Solubility in water: .. Other solubilities: soluble in ether,thf,dmso,dmf,methanol | |
B(C1=C(C=C(C=C1)F)F)(O)O | |
157.91 | |
157.91 |
144025-03-6 | |
100.0 | |
97.5% min. (HPLC) | |
C6H5BF2O2 | |
2,4-difluorobenzeneboronic acid, 2,4-difluorophenyl boronic acid, 2,4-fluorobenzene boronic acid, 2,4-difluorophenyl boranediol, 2,4-difluorophenyl-boronic acid, 2,4-difluorophenylboronicacid, 2 4-difluorophenylboronic acid, boronic acid, 2,4-difluorophenyl, boronic acid, b-2,4-difluorophenyl | |
QQLRSCZSKQTFGY-UHFFFAOYSA-N | |
(2,4-difluorophenyl)boronic acid | |
2734334 | |
98% |
Chemical Identifiers
144025-03-6 | |
157.91 | |
2,4-difluorobenzeneboronic acid, 2,4-difluorophenyl boronic acid, 2,4-fluorobenzene boronic acid, 2,4-difluorophenyl boranediol, 2,4-difluorophenyl-boronic acid, 2,4-difluorophenylboronicacid, 2 4-difluorophenylboronic acid, boronic acid, 2,4-difluorophenyl, boronic acid, b-2,4-difluorophenyl | |
(2,4-difluorophenyl)boronic acid |
C6H5BF2O2 | |
QQLRSCZSKQTFGY-UHFFFAOYSA-N | |
2734334 | |
B(C1=C(C=C(C=C1)F)F)(O)O |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER
GHS Signal Word: Warning
RUO â Research Use Only