missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Difluorophenylacetic acid, 99%

$163.16 - $631.11

Chemical Identifiers

CAS 81228-09-3
Molecular Formula C8H6F2O2
Formula Weight 172.13
MDL Number MFCD00009999
Beilstein 3649727
Chemical Name or Material 2,4-Difluorophenylacetic acid
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2043106 SDS Alfa Aesar™
B2043106
5g Each for $163.16
AAB2043114 SDS Alfa Aesar™
B2043114
25g Each for $631.11
Specifications Description & Specifications

Chemical Identifiers

81228-09-3
172.13
3649727
C8H6F2O2
MFCD00009999
2,4-Difluorophenylacetic acid

Specifications

117° to 119°C
81228-09-3
172.13
3649727
99%
5g
C8H6F2O2
MFCD00009999
2,4-Difluorophenylacetic acid

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning