Please login to your online account to display your discounted pricing

2,4-Difluorophenylacetic acid, 97%, ACROS Organics™

$42.64 - $42.64

Chemical Identifiers

Chemical Name or Material 2, 4-Difluorophenylacetic acid
CAS 81228-09-3
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C8H6F2O2
Linear Formula F2C6H3CH2CO2H
Formula Weight 172.13
MDL Number MFCD00009999
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC303550010

  SDS
thermo fisher scientific
303550010
Glass bottle 1g Each for $42.64
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Powder
Formula Weight 172.13
Quantity 1g
Color White
Melting Point 115° to 120°C
CAS Min % 96.0
CAS Max % 100.0
Molecular Formula C8H6F2O2
Linear Formula F2C6H3CH2CO2H
CAS 81228-09-3
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Chemical Name or Material 2, 4-Difluorophenylacetic acid
Infrared Spectrum Authentic
Assay Percent Range 97%
MDL Number MFCD00009999
Packaging Glass bottle

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.