Please login to your online account to display your discounted pricing

2,4-Difluorophenol, 99+%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2, 4-Difluorophenol
Name Note 99+%
CAS 367-27-1
CAS Min % 99.0
CAS Max % 100.0
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C6H4F2O
Linear Formula F2(C6H3)OH
Formula Weight 130.09
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC243200100

  SDS
thermo fisher scientific
243200100
Glass bottle 10mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥99.0%
Physical Form Liquid After Melting
Formula Weight 130.09
Quantity 10mL
Color Clear yellow-green
EINECS Number 206-688-0
TSCA TSCA
Density 1.3600g/mL
CAS Min % 99.0
CAS Max % 100.0
Chemical Name or Material 2, 4-Difluorophenol
Infrared Spectrum Authentic
Name Note 99+%
Assay Percent Range 99+%
Molecular Formula C6H4F2O
Linear Formula F2(C6H3)OH
MDL Number MFCD00009715
CAS 367-27-1
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Flammable solid.
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep container tightly closed.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Refractive Index 1.4840 to 1.4860 (20°C, 589nm)
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep container tightly closed.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Flammable solid.