Please login to your online account to display your discounted pricing

2,4-Difluoronitrobenzene, 99%, ACROS Organics™

$17.71 - $17.71

Chemical Identifiers

Chemical Name or Material 2, 4-Difluoronitrobenzene
CAS 446-35-5
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C6H3F2NO2
Linear Formula F2C6H3NO2
Formula Weight 159.09
MDL Number MFCD00007050
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC214480050

  SDS
thermo fisher scientific
214480050
Glass bottle 5mL Each for $17.71
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Liquid
Color Clear yellow to brownish
Formula Weight 159.09
Quantity 5mL
Density 1.4500g/mL
Beilstein 05, I, 129
CAS Max % 100.0
Molecular Formula C6H3F2NO2
Packaging Glass bottle
Assay Percent Range 99%
CAS 446-35-5
Health Hazard 1 GHS Signal Word: Danger
CAS Min % 98.5
MDL Number MFCD00007050
Linear Formula F2C6H3NO2
Refractive Index 1.5110 to 1.5130 (20°C, 589nm)
Infrared Spectrum Authentic
Chemical Name or Material 2, 4-Difluoronitrobenzene
Health Hazard 2 GHS H Statement:
Toxic if swallowed.
Toxic in contact with skin.
Causes serious eye irritation.
Toxic if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic if swallowed.
Toxic in contact with skin.
Causes serious eye irritation.
Toxic if inhaled.
May cause respiratory irritation.
Causes skin irritation.