Please login to your online account to display your discounted pricing

2,4-Difluoronitrobenzene, 99%, ACROS Organics™


Quantity 5mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  214480050

 View more versions of this product

Catalog No. AC214480050

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Liquid
Formula Weight 159.09
Color Clear yellow to brownish
Quantity 5mL
Density 1.4500g/mL
Beilstein 05, I, 129
CAS Max % 100.0
Assay Percent Range 99%
Molecular Formula C6H3F2NO2
CAS 446-35-5
Packaging Glass bottle
Linear Formula F2C6H3NO2
MDL Number MFCD00007050
CAS Min % 98.5
Health Hazard 1 GHS Signal Word: Danger
Chemical Name or Material 2, 4-Difluoronitrobenzene
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Toxic if swallowed.
Toxic in contact with skin.
Causes serious eye irritation.
Toxic if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Refractive Index 1.5110 to 1.5130 (20°C, 589nm)

Specifications

Chemical Name or Material 2, 4-Difluoronitrobenzene
CAS 446-35-5
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C6H3F2NO2
Linear Formula F2C6H3NO2
Formula Weight 159.09
MDL Number MFCD00007050
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic if swallowed.
Toxic in contact with skin.
Causes serious eye irritation.
Toxic if inhaled.
May cause respiratory irritation.
Causes skin irritation.

SDS & Safety Documents