missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,4-Difluorobenzylamine, 98%, Thermo Scientific™

Manufacturer:  Thermo Fisher Scientific 305660010

 View more versions of this product

Catalog No. AC305660010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

2, 4-Difluorobenzylamine
97.5
1.2000g/mL
82°C to 84°C (15.0mmHg)
97.5% min. (GC)
C7H7F2N
MFCD00010142
1.4900 to 1.493
1g
1.2
(2,4-difluorophenyl)methanamine
2733244
Liquid
72235-52-0
100.0
Colorless to Yellow
68°C
Authentic
F2C6H3CH2NH2
Glass bottle
1-2,4-difluorophenyl methanamine, 2,4-difluorbenzylamine, 2,4-difluorobenzyl amine, 2,4-difluorobenzylamine, 2,4-difluorophenyl methanamine, 2,4-difluorophenyl methylamine, acmc-209oma, benzenemethanamine, 2,4-difluoro, n-difluorobenzylamine, pubchem4399
QDZZDVQGBKTLHV-UHFFFAOYSA-N
C1=CC(=C(C=C1F)F)CN
143.14
143.14
98%

Chemical Identifiers

72235-52-0
143.14
QDZZDVQGBKTLHV-UHFFFAOYSA-N
2733244
C1=CC(=C(C=C1F)F)CN
C7H7F2N
MFCD00010142
1-2,4-difluorophenyl methanamine, 2,4-difluorbenzylamine, 2,4-difluorobenzyl amine, 2,4-difluorobenzylamine, 2,4-difluorophenyl methanamine, 2,4-difluorophenyl methylamine, acmc-209oma, benzenemethanamine, 2,4-difluoro, n-difluorobenzylamine, pubchem4399
(2,4-difluorophenyl)methanamine
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 276-502-

SDS Documents
Product Certifications
Promotions

Promotions

RUO – Research Use Only