Please login to your online account to display your discounted pricing

2,4-Difluorobenzylamine, 98%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC305660050

 View more versions of this product

Catalog No. AC305660050

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Liquid
Formula Weight 143.14
Quantity 5g
Color Clear colorless to light yellow
Density 1.2g/mL
CAS Max % 100.0
Health Hazard 1 GHS Signal Word: Danger
Linear Formula FC6H3CHNH
Assay Percent Range 98%
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS Min % 97.5
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4900 to 1.4930 (20°C, 589nm)
Chemical Name or Material 2, 4-Difluorobenzylamine
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
CAS 72235-52-0
Molecular Formula C7H7F2N
MDL Number MFCD00010142

Specifications

Chemical Name or Material 2, 4-Difluorobenzylamine
CAS 72235-52-0
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C7H7F2N
Linear Formula FC6H3CHNH
Formula Weight 143.14
MDL Number MFCD00010142
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

SDS & Safety Documents