Learn More
2,4-Difluorobenzyl bromide, 98+%, Thermo Scientific Chemicals
$112.16 - $351.03
Chemical Identifiers
CAS | 23915-07-3 |
---|---|
Molecular Formula | C7H5BrF2 |
Molecular Weight (g/mol) | 207.018 |
MDL Number | MFCD00011649 |
InChI Key | IBLMYGXJKQIGSN-UHFFFAOYSA-N |
Synonym | 2,4-difluorobenzyl bromide, 1-bromomethyl-2,4-difluorobenzene, 2,4-difluorobenzylbromide, alpha-bromo-2,4-difluorotoluene, benzene, 1-bromomethyl-2,4-difluoro, 2,4-difluoro benzyl bromide, 1-bromomethyl-2,4-difluoro-benzene, 2-chloromethyl quinoline hcl salt, fr cf d1e, pubchem4911 |
PubChem CID | 90297 |
IUPAC Name | 1-(bromomethyl)-2,4-difluorobenzene |
SMILES | C1=CC(=C(C=C1F)F)CBr |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1521306
|
Thermo Scientific Chemicals
A1521306 |
5 g |
Each for $112.16
|
|
|||||
AAA1521314
|
Thermo Scientific Chemicals
A1521314 |
25 g |
Each for $351.03
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
23915-07-3 | |
207.018 | |
IBLMYGXJKQIGSN-UHFFFAOYSA-N | |
90297 | |
C1=CC(=C(C=C1F)F)CBr |
C7H5BrF2 | |
MFCD00011649 | |
2,4-difluorobenzyl bromide, 1-bromomethyl-2,4-difluorobenzene, 2,4-difluorobenzylbromide, alpha-bromo-2,4-difluorotoluene, benzene, 1-bromomethyl-2,4-difluoro, 2,4-difluoro benzyl bromide, 1-bromomethyl-2,4-difluoro-benzene, 2-chloromethyl quinoline hcl salt, fr cf d1e, pubchem4911 | |
1-(bromomethyl)-2,4-difluorobenzene |
Specifications
23915-07-3 | |
42°C to 43°C (1 mmHg) | |
C7H5BrF2 | |
MFCD00011649 | |
UN1760 | |
2,4-difluorobenzyl bromide, 1-bromomethyl-2,4-difluorobenzene, 2,4-difluorobenzylbromide, alpha-bromo-2,4-difluorotoluene, benzene, 1-bromomethyl-2,4-difluoro, 2,4-difluoro benzyl bromide, 1-bromomethyl-2,4-difluoro-benzene, 2-chloromethyl quinoline hcl salt, fr cf d1e, pubchem4911 | |
C1=CC(=C(C=C1F)F)CBr | |
207.018 | |
207.02 | |
2,4-Difluorobenzyl bromide |
1.613 | |
85°C (185°F) | |
1.525 | |
5 g | |
4177539 | |
IBLMYGXJKQIGSN-UHFFFAOYSA-N | |
1-(bromomethyl)-2,4-difluorobenzene | |
90297 | |
≥98% |
Safety and Handling
GHS H Statement
H314-H318-H317-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause an allergic skin reaction.
Combustible liquid.
P210-P233-P240-P241-P242-P243-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H226-H314
DOTInformation : Hazard Class: 3; Packaging Group: III
EINECSNumber : 245-938-3
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only