Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Difluorobenzyl alcohol, 98+%

$163.16 - $163.16

Chemical Identifiers

Chemical Name or Material 2,4-Difluorobenzyl alcohol
CAS 56456-47-4
Molecular Formula C7H6F2O
Formula Weight 144.12
MDL Number MFCD00009983
Beilstein 2501944
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1994509

 
alfa aesar™
A1994509
10g Each for $163.16
Description & Specifications

Specifications

Formula Weight 144.12
Chemical Name or Material 2,4-Difluorobenzyl alcohol
Assay ≥98%
Molecular Formula C7H6F2O
MDL Number MFCD00009983
CAS 56456-47-4
Beilstein 2501944
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Flash Point 90°C (194°F)
Boiling Point 51° to 52°C (1mmHg)
Quantity 10g
Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.