Please login to your online account to display your discounted pricing

2,4-Difluorobenzoyl chloride, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2, 4-Difluorobenzoyl chloride
CAS 72482-64-5
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C7H3ClF2O
Linear Formula F2C6H3COCl
Formula Weight 176.55
MDL Number MFCD00000658
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC297220025

  SDS
thermo fisher scientific
297220025
Glass bottle 2.5mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC297220100

  SDS
thermo fisher scientific
297220100
Glass bottle 10mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Liquid
Formula Weight 176.55
Quantity 2.5mL
Color Clear light yellow to yellow
Density 1.42g/mL
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material 2, 4-Difluorobenzoyl chloride
Refractive Index 1.5147 to 1.5167 (20°C, 589nm)
Assay Percent Range 97%
Molecular Formula C7H3ClF2O
Linear Formula F2C6H3COCl
CAS 72482-64-5
Packaging Glass bottle
MDL Number MFCD00000658
Health Hazard 1 GHS Signal Word: Danger
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.