Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Difluorobenzophenone, 97+%

$130.47 - $130.47

Chemical Identifiers

Chemical Name or Material 2,4-Difluorobenzophenone
CAS 85068-35-5
Molecular Formula C13H8F2O
Formula Weight 218.2
MDL Number MFCD00000331
Beilstein 6596192
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2304606

  SDS
alfa aesar™
B2304606
5g Each for $130.47
Description & Specifications

Specifications

Formula Weight 218.2
Quantity 5g
Chemical Name or Material 2,4-Difluorobenzophenone
Health Hazard 1 Warning
Boiling Point 119° to 120°C (2mmHg)
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Assay ≥97%
MDL Number MFCD00000331
Beilstein 6596192
CAS 85068-35-5
Molecular Formula C13H8F2O
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Flash Point >110°C (230°F)
Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.