Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4'-Difluorobenzophenone, 97%

$34.66 - $34.66

Chemical Identifiers

Chemical Name or Material 2,4'-Difluorobenzophenone
CAS 342-25-6
Molecular Formula C13H8F2O
Formula Weight 218.2
MDL Number MFCD00000319
Beilstein 3268520
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1030606

  SDS
alfa aesar™
L1030606
5g Each for $34.66
Description & Specifications

Specifications

Formula Weight 218.2
Chemical Name or Material 2,4'-Difluorobenzophenone
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Assay 97%
CAS 342-25-6
MDL Number MFCD00000319
Health Hazard 1 Warning
Flash Point >110°C (230°F)
Molecular Formula C13H8F2O
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Beilstein 3268520
Quantity 5g
Boiling Point 176° to 178°C (16mmHg)
Melting Point 22° to 24°C
Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.