missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Difluorobenzenesulfonyl chloride, 98%

Manufacturer:  Alfa Aesar™ A1463906

 View more versions of this product

Catalog No. AAA1463906



Specifications

Specifications

2,4-Difluorobenzenesulfonyl chloride
236°C to 237°C
13918-92-8
MFCD00042481
2970373
FJSAJUXIHJIAMD-UHFFFAOYSA-N
2,4-difluorobenzenesulfonyl chloride
2734273
98%
>110°C (230°F)
5g
C6H3ClF2O2S
UN3265
2,4-difluorobenzene-1-sulfonyl chloride, 2,4-difluorobenzenesulphonyl chloride, 2,4-difluoro-benzenesulfonyl chloride, benzenesulfonyl chloride, 2,4-difluoro, 2,4-difluorophenyl chlorosulfone, buttpark 23\07-79, 2,4-difluorophenylsulfonyl chloride, 2,4-difluorobenzenesulphonyl chloride, pubchem5122, acmc-209civ
C1=CC(=C(C=C1F)F)S(=O)(=O)Cl
212.595
212.6

Chemical Identifiers

13918-92-8
212.595
FJSAJUXIHJIAMD-UHFFFAOYSA-N
2734273
C1=CC(=C(C=C1F)F)S(=O)(=O)Cl
C6H3ClF2O2S
MFCD00042481
2,4-difluorobenzene-1-sulfonyl chloride, 2,4-difluorobenzenesulphonyl chloride, 2,4-difluoro-benzenesulfonyl chloride, benzenesulfonyl chloride, 2,4-difluoro, 2,4-difluorophenyl chlorosulfone, buttpark 23\07-79, 2,4-difluorophenylsulfonyl chloride, 2,4-difluorobenzenesulphonyl chloride, pubchem5122, acmc-209civ
2,4-difluorobenzenesulfonyl chloride
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

Documents