missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Difluorobenzeneboronic acid 1,3-propanediol ester, 98%

Manufacturer:  Alfa Aesar™ H6152503

 View more versions of this product

Catalog No. AAH6152503



Specifications

Specifications

2,4-Difluorobenzeneboronic acid 1,3-propanediol ester
1073372-07-2
MFCD11504975
AHKNGRYTXDKKDQ-UHFFFAOYSA-N
2-(2,4-difluorophenyl)-1,3,2-dioxaborinane
46739109
Solid
1g
C9H9BF2O2
2,4-difluorobenzeneboronic acid 1,3-propanediol ester
B1(OCCCO1)C2=C(C=C(C=C2)F)F
197.976
197.98
98%

Chemical Identifiers

1073372-07-2
197.976
AHKNGRYTXDKKDQ-UHFFFAOYSA-N
46739109
B1(OCCCO1)C2=C(C=C(C=C2)F)F
C9H9BF2O2
MFCD11504975
2,4-difluorobenzeneboronic acid 1,3-propanediol ester
2-(2,4-difluorophenyl)-1,3,2-dioxaborinane
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

Documents