Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Difluorobenzeneboronic acid 1,3-propanediol ester, 98%

Chemical Identifiers

Chemical Name or Material 2,4-Difluorobenzeneboronic acid 1,3-propanediol ester
CAS 1073372-07-2
Formula Weight 197.98
MDL Number MFCD11504975
Synonym 2-(2,4-Difluorophenyl)-1,3,2-dioxaborinane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6152503

 
alfa aesar™
H6152503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6152506

 
alfa aesar™
H6152506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 197.98
Physical Form Solid
MDL Number MFCD11504975
Chemical Name or Material 2,4-Difluorobenzeneboronic acid 1,3-propanediol ester
Assay 98%
CAS 1073372-07-2
Synonym 2-(2,4-Difluorophenyl)-1,3,2-dioxaborinane
Quantity 1g