Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Difluorobenzamidoxime, 99%

$353.59 - $353.59

Chemical Identifiers

Chemical Name or Material 2,4-Difluorobenzamidoxime
CAS 883022-90-0
Molecular Formula C7H6F2N2O
Formula Weight 172.13g/mol
MDL Number MFCD05662977
Synonym 2,4-Difluoro-N-hydroxybenzenecarboximidamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5066403

 
alfa aesar™
H50664-03
Each for $353.59
Description & Specifications

Specifications

Quantity 1g
Chemical Name or Material 2,4-Difluorobenzamidoxime
CAS 883022-90-0
Assay 99%
Molecular Formula C7H6F2N2O
Formula Weight 172.13g/mol
MDL Number MFCD05662977
Synonym 2,4-Difluoro-N-hydroxybenzenecarboximidamide
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.