missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Difluorobenzamide, 97%

$51.20 - $168.09

Chemical Identifiers

CAS 85118-02-1
Molecular Formula C7H5F2NO
Molecular Weight (g/mol) 157.12
MDL Number MFCD00015547
InChI Key KTXFXDMDYZIXSJ-UHFFFAOYSA-N
Synonym benzamide, 2,4-difluoro, difluoro benzamide, pubchem3458, acmc-209q4l, 2,4-difluorobenzamide, 2,4-bis fluoranyl benzamide, 2,4-difluorobenzamide
PubChem CID 123588
IUPAC Name 2,4-difluorobenzamide
SMILES C1=CC(=C(C=C1F)F)C(=O)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1037506 View Documents Alfa Aesar™
A1037506
5g Each for $168.09
 
AAA1037503 View Documents Alfa Aesar™
A1037503
1g Each for $51.20
 
Specifications

Chemical Identifiers

85118-02-1
157.12
KTXFXDMDYZIXSJ-UHFFFAOYSA-N
123588
C1=CC(=C(C=C1F)F)C(=O)N
C7H5F2NO
MFCD00015547
benzamide, 2,4-difluoro, difluoro benzamide, pubchem3458, acmc-209q4l, 2,4-difluorobenzamide, 2,4-bis fluoranyl benzamide, 2,4-difluorobenzamide
2,4-difluorobenzamide

Specifications

160°C to 162°C
85118-02-1
MFCD00015547
KTXFXDMDYZIXSJ-UHFFFAOYSA-N
2,4-difluorobenzamide
123588
97%
5g
C7H5F2NO
benzamide, 2,4-difluoro, difluoro benzamide, pubchem3458, acmc-209q4l, 2,4-difluorobenzamide, 2,4-bis fluoranyl benzamide, 2,4-difluorobenzamide
C1=CC(=C(C=C1F)F)C(=O)N
157.12
157.12
2,4-Difluorobenzamide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning