Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Difluoroanisole, 98%

Chemical Identifiers

Chemical Name or Material 2,4-Difluoroanisole
CAS 452-10-8
Molecular Formula C7H6F2O
Formula Weight 144.12
MDL Number MFCD00010731
UN Number UN1993
Beilstein 2518264
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2150906

 
alfa aesar™
B2150906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2150914

 
alfa aesar™
B2150914
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 144.12
UN Number UN1993
Health Hazard 2 GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flash Point 51°C (123°F)
Chemical Name or Material 2,4-Difluoroanisole
Assay 98%
Molecular Formula C7H6F2O
MDL Number MFCD00010731
CAS 452-10-8
Beilstein 2518264
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Boiling Point 53°C (18mmHg)
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.