missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,4-Difluoroaniline, 99%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC114591000
Specifications
2, 4-Difluoroaniline | |
62°C | |
Glass bottle | |
1.268 | |
Brown | |
100mL | |
98.5 | |
98.5% min. (GC) | |
F2C6H3NH2 | |
12, I, 297 | |
1-amino-2,4-difluorobenzene, 2,4-difluoro aniline, 2,4-difluoro-phenylamine, 2,4-difluorobenzenamine, 2,4-difluorophenylamine, 2,4-difluroaniline, aniline, 2,4-difluoro, benzenamine, 2,4-difluoro, ccris 4621, unii-40p93l7kwd | |
CEPCPXLLFXPZGW-UHFFFAOYSA-N | |
2,4-difluoroaniline | |
9709 | |
Liquid |
1.2680g/mL | |
Authentic | |
1.5025 to 1.5075 | |
170°C (753.0mmHg) | |
-7.5°C | |
367-25-9 | |
100.0 | |
C6H5F2N | |
MFCD00007648 | |
15, 3166 | |
Solubility in water: immiscible | |
C1=CC(=C(C=C1F)F)N | |
129.11 | |
129.11 | |
99% |
Chemical Identifiers
367-25-9 | |
129.11 | |
CEPCPXLLFXPZGW-UHFFFAOYSA-N | |
9709 | |
C1=CC(=C(C=C1F)F)N |
C6H5F2N | |
MFCD00007648 | |
1-amino-2,4-difluorobenzene, 2,4-difluoro aniline, 2,4-difluoro-phenylamine, 2,4-difluorobenzenamine, 2,4-difluorophenylamine, 2,4-difluroaniline, aniline, 2,4-difluoro, benzenamine, 2,4-difluoro, ccris 4621, unii-40p93l7kwd | |
2,4-difluoroaniline |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfort
GHS Signal Word: Danger
EINECSNumber : 206-687-5
Recommended Storage : Refrigerator +4°C