Please login to your online account to display your discounted pricing

Alfa Aesar™ 2',4'-Difluoroacetophenone, 98%

$85.74 - $85.74

Chemical Identifiers

Chemical Name or Material 2',4'-Difluoroacetophenone
CAS 364-83-0
Molecular Formula C8H6F2O
Formula Weight 156.13
MDL Number MFCD00151261
Beilstein 1940741
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1342006

  SDS
alfa aesar™
A1342006
5g Each for $85.74
Description & Specifications

Specifications

Formula Weight 156.13
Chemical Name or Material 2',4'-Difluoroacetophenone
TSCA TSCA
Assay 98%
Molecular Formula C8H6F2O
MDL Number MFCD00151261
CAS 364-83-0
Beilstein 1940741
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Flash Point 66°C (150°F)
Odor Characteristic
Boiling Point 80° to 81°C (25mmHg)
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.