missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2',4'-Difluoroacetanilide, 98%

$38.09 - $482.69

Chemical Identifiers

CAS 399-36-0
Molecular Formula C8H7F2NO
Molecular Weight (g/mol) 171.147
MDL Number MFCD00032502
InChI Key WOHLPEUHFSHZAN-UHFFFAOYSA-N
Synonym 2',4'-difluoroacetanilide, n-2,4-difluorophenyl acetamide, 2,4-difluoroacetanilide, 2,4-difluorophenyl acetamide, acetanilide,2,4-difluoro, acetamide, n-2,4-difluorophenyl, acetamide,n-2,4-difluorophenyl, aminobenzene, n-acetyl-2,4-difluoro, acetanilide, 2',4'-difluoro, maybridge1_001023
PubChem CID 96093
IUPAC Name N-(2,4-difluorophenyl)acetamide
SMILES CC(=O)NC1=C(C=C(C=C1)F)F
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1049014 View Documents Alfa Aesar™
A1049014
25g Each for $38.09
 
AAA1049036 View Documents Alfa Aesar™
A1049036
500g Each for $482.69
 
AAA1049022 View Documents Alfa Aesar™
A1049022
100g Each for $115.04
 
Specifications

Chemical Identifiers

399-36-0
171.147
WOHLPEUHFSHZAN-UHFFFAOYSA-N
96093
CC(=O)NC1=C(C=C(C=C1)F)F
C8H7F2NO
MFCD00032502
2',4'-difluoroacetanilide, n-2,4-difluorophenyl acetamide, 2,4-difluoroacetanilide, 2,4-difluorophenyl acetamide, acetanilide,2,4-difluoro, acetamide, n-2,4-difluorophenyl, acetamide,n-2,4-difluorophenyl, aminobenzene, n-acetyl-2,4-difluoro, acetanilide, 2',4'-difluoro, maybridge1_001023
N-(2,4-difluorophenyl)acetamide

Specifications

120°C to 123°C
399-36-0
MFCD00032502
2',4'-difluoroacetanilide, n-2,4-difluorophenyl acetamide, 2,4-difluoroacetanilide, 2,4-difluorophenyl acetamide, acetanilide,2,4-difluoro, acetamide, n-2,4-difluorophenyl, acetamide,n-2,4-difluorophenyl, aminobenzene, n-acetyl-2,4-difluoro, acetanilide, 2',4'-difluoro, maybridge1_001023
CC(=O)NC1=C(C=C(C=C1)F)F
171.147
171.15
2',4'-Difluoroacetanilide
25g
C8H7F2NO
2832300
WOHLPEUHFSHZAN-UHFFFAOYSA-N
N-(2,4-difluorophenyl)acetamide
96093
98%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning