Please login to your online account to display your discounted pricing

Alfa Aesar™ 2',4'-Difluoro-N-methylacetanilide, 97%

$42.19 - $564.92

Chemical Identifiers

Chemical Name or Material 2',4'-Difluoro-N-methylacetanilide
CAS 238403-47-9
Molecular Formula C9H9F2NO
Formula Weight 185.17
MDL Number MFCD01567301
Synonym N-(2,4-Difluorophenyl)-N-methylacetamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2493503

  SDS
alfa aesar™
B2493503
1g Each for $42.19

AAB2493506

  SDS
alfa aesar™
B2493506
5g Each for $149.31

AAB2493514

  SDS
alfa aesar™
B2493514
25g Each for $564.92
Description & Specifications

Specifications

Formula Weight 185.17
Chemical Name or Material 2',4'-Difluoro-N-methylacetanilide
Assay 97%
Molecular Formula C9H9F2NO
MDL Number MFCD01567301
CAS 238403-47-9
Synonym N-(2,4-Difluorophenyl)-N-methylacetamide
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 83° to 85°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.