missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,4-Difluoro-3-nitrobenzonitrile, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC451420050
Specifications
2, 4-Difluoro-3-nitrobenzonitrile | |
5g | |
96% min. (GC) | |
3-nitro-2,4-difluoro-benzonitrile | |
C1=CC(=C(C(=C1C#N)F)[N+](=O)[O-])F | |
184.1 | |
184.1 |
1.529 | |
1186194-75-1 | |
C7H2F2N2O2 | |
HESWWHRCQMLPFT-UHFFFAOYSA-N | |
2,4-difluoro-3-nitrobenzonitrile | |
45790497 | |
97% |
Chemical Identifiers
1186194-75-1 | |
184.1 | |
3-nitro-2,4-difluoro-benzonitrile | |
2,4-difluoro-3-nitrobenzonitrile |
C7H2F2N2O2 | |
HESWWHRCQMLPFT-UHFFFAOYSA-N | |
45790497 | |
C1=CC(=C(C(=C1C#N)F)[N+](=O)[O-])F |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of
GHS Signal Word: Danger