Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Difluoro-3-methylbenzeneboronic acid pinacol ester, 96%

Chemical Identifiers

Chemical Name or Material 2,4-Difluoro-3-methylbenzeneboronic acid pinacol ester
Formula Weight 254.08
Synonym 2,4-Difluoro-3-methylphenylboronic acid pinacol ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6288403

 
alfa aesar™
H6288403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH62884MD

 
alfa aesar™
H62884MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 254.08
Synonym 2,4-Difluoro-3-methylphenylboronic acid pinacol ester
Chemical Name or Material 2,4-Difluoro-3-methylbenzeneboronic acid pinacol ester
Assay 96%
Quantity 1g
Physical Form Solid