Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Difluoro-3-methoxyphenol, 97%

$200.41 - $611.74

Chemical Identifiers

Chemical Name or Material 2,4-Difluoro-3-methoxyphenol
CAS 886499-27-0
Molecular Formula C7H6F2O2
Formula Weight 160.12
MDL Number MFCD04115938
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2637103

  SDS
alfa aesar™
H2637103
1g Each for $200.41

AAH2637106

  SDS
alfa aesar™
H2637106
5g Each for $611.74
Description & Specifications

Specifications

Formula Weight 160.12
Chemical Name or Material 2,4-Difluoro-3-methoxyphenol
Assay 97%
Molecular Formula C7H6F2O2
MDL Number MFCD04115938
CAS 886499-27-0
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H318-H302-H335-H315
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF eye irritation persists: Get medical advice/attention.
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF eye irritation persists: Get medical advice/attention.

GHS H Statement
H318-H302-H335-H315
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.