Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Difluoro-3-methoxybenzaldehyde, 97%

$153.69 - $860.36

Chemical Identifiers

Chemical Name or Material 2,4-Difluoro-3-methoxybenzaldehyde
CAS 870837-66-4
Molecular Formula C8H6F2O2
Formula Weight 172.13
MDL Number MFCD04115930
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2659703

  SDS
alfa aesar™
H2659703
1g Each for $153.69

AAH2659709

  SDS
alfa aesar™
H2659709
10g Each for $860.36
Description & Specifications

Specifications

Formula Weight 172.13
Chemical Name or Material 2,4-Difluoro-3-methoxybenzaldehyde
Assay 97%
Molecular Formula C8H6F2O2
MDL Number MFCD04115930
CAS 870837-66-4
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g

Applications
2,4-Difluoro-3-methoxybenzaldehyde is used as a laboratory reagent. It is also used as an intermediate in pharmaceutical industries.

Solubility
Insoluble in water.

Notes
Air sensitive. Incompatible with strong oxidizing agents, strong acids and bases. Store in a cool place.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.