missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Difluoro-3-iodopyridine, 95%

$141.42 - $422.54

Chemical Identifiers

CAS 837364-88-2
Molecular Formula C5H2F2IN
Molecular Weight (g/mol) 240.979
MDL Number MFCD09029861
InChI Key AYKASHJPLZSRDC-UHFFFAOYSA-N
Synonym 2,4-difluoro-3-iodo-pyridine, pyridine, 2,4-difluoro-3-iodo, 2,4-difluoroi-3-iodo-pyridine
PubChem CID 12175804
IUPAC Name 2,4-difluoro-3-iodopyridine
SMILES C1=CN=C(C(=C1F)I)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH66280MD View Documents Alfa Aesar™
H66280MD
250mg Each for $141.42
 
AAH6628003 View Documents Alfa Aesar™
H6628003
1g Each for $422.54
 
Specifications

Chemical Identifiers

837364-88-2
240.979
AYKASHJPLZSRDC-UHFFFAOYSA-N
12175804
C1=CN=C(C(=C1F)I)F
C5H2F2IN
MFCD09029861
2,4-difluoro-3-iodo-pyridine, pyridine, 2,4-difluoro-3-iodo, 2,4-difluoroi-3-iodo-pyridine
2,4-difluoro-3-iodopyridine

Specifications

Light Sensitive
837364-88-2
MFCD09029861
2,4-difluoro-3-iodo-pyridine, pyridine, 2,4-difluoro-3-iodo, 2,4-difluoroi-3-iodo-pyridine
C1=CN=C(C(=C1F)I)F
240.979
240.98
95%
250mg
C5H2F2IN
9995192
AYKASHJPLZSRDC-UHFFFAOYSA-N
2,4-difluoro-3-iodopyridine
12175804
Solid
2,4-Difluoro-3-iodopyridine
Safety and Handling

Safety and Handling

storageNote1 : -30° to -10°C