missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Dichlorothiophenol, 97%

Manufacturer:  Alfa Aesar™ A1238403

 View more versions of this product

Catalog No. AAA1238403



Specifications

Specifications

2,4-Dichlorothiophenol
Unpleasant
~15°C
1122-41-4
C6H4Cl2S
2042868
FGBVJFREPSJSNG-UHFFFAOYSA-N
2,4-dichlorobenzenethiol
70730
>110°C (230°F)
60°C to 62°C (1.5mmHg)
1g
97%
MFCD00041421
2,4-Dichlorobenzenethiol
C1=CC(=C(C=C1Cl)Cl)S
179.058
179.07

Chemical Identifiers

1122-41-4
179.058
FGBVJFREPSJSNG-UHFFFAOYSA-N
70730
C1=CC(=C(C=C1Cl)Cl)S
C6H4Cl2S
MFCD00041421
2,4-Dichlorobenzenethiol
2,4-dichlorobenzenethiol
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280h-P305+P351+P338
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.

Warning

SDS Documents