Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Dichlorothiazole-5-carboxaldehyde, 96%

$127.18 - $439.97

Chemical Identifiers

Chemical Name or Material 2,4-Dichlorothiazole-5-carboxaldehyde
CAS 92972-48-0
Molecular Formula C4HCl2NOS
Formula Weight 182.03
MDL Number MFCD00793013
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5045603

  SDS
alfa aesar™
H5045603
1g Each for $439.97

AAH50456MD

  SDS
alfa aesar™
H50456MD
250mg Each for $127.18
Description & Specifications

Specifications

Formula Weight 182.03
Chemical Name or Material 2,4-Dichlorothiazole-5-carboxaldehyde
Assay 96%
Molecular Formula C4HCl2NOS
MDL Number MFCD00793013
CAS 92972-48-0
Melting Point 48° to 49°C
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.