missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Dichloropyrimidine, 98+%

Manufacturer:  Alfa Aesar™ A1513106

 View more versions of this product

Catalog No. AAA1513106



Description

Description

Applications
2,4-Dichloropyrimidine is used in the synthesis of medicinally important 4-aryl-5-pyrimidinylimidazoles. It undergoes effective one-pot, regioselective double Suzuki coupling reaction to yield diarylated pyrimidines.

Solubility
Soluble in water (partly), methanol, chloroform, and ethyl acetate.

Notes
Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

2,4-Dichloropyrimidine
57°C to 62°C
3934-20-1
MFCD00006061
pyrimidine, 2,4-dichloro, 2,6-dichloropyrimidine, 2,4-dichloro-pyrimidine, 2,4-dichlorpyrimidin, 2,4-dichloro pyrimidine, pyrimidine,2,4-dichloro, pyrimidine,4-dichloro, hydrochloride, zlchem 210, 2,4dichloropyrimidine
C1=CN=C(N=C1Cl)Cl
148.974
148.98
101°C (23mmHg)
5g
C4H2Cl2N2
110911
BTTNYQZNBZNDOR-UHFFFAOYSA-N
2,4-dichloropyrimidine
77531
≥98%

Chemical Identifiers

3934-20-1
148.974
BTTNYQZNBZNDOR-UHFFFAOYSA-N
77531
C1=CN=C(N=C1Cl)Cl
C4H2Cl2N2
MFCD00006061
pyrimidine, 2,4-dichloro, 2,6-dichloropyrimidine, 2,4-dichloro-pyrimidine, 2,4-dichlorpyrimidin, 2,4-dichloro pyrimidine, pyrimidine,2,4-dichloro, pyrimidine,4-dichloro, hydrochloride, zlchem 210, 2,4dichloropyrimidine
2,4-dichloropyrimidine
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

Recommended Storage : 2° to 8°C

Documents