Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Dichloropyridine, 97%

Chemical Identifiers

Chemical Name or Material 2,4-Dichloropyridine
CAS 26452-80-2
Molecular Formula C5H3Cl2N
Formula Weight 147.99
MDL Number MFCD00955618
UN Number UN2810
Beilstein 108666
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL2004003

  SDS
alfa aesar™
L2004003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL2004006

  SDS
alfa aesar™
L2004006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 147.99
MDL Number MFCD00955618
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338-P361-P309+P311-P302+P352
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
IF exposed or if you feel unwell: Call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Quantity 1g
CAS 26452-80-2
Molecular Formula C5H3Cl2N
Health Hazard 2 GHS H Statement
H311-H227-H302-H315-H319-H335
Toxic in contact with skin.
Combustible liquid.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Boiling Point 189° to 190°C
Health Hazard 1 Danger
Assay 97%
Melting Point −1°C
UN Number UN2810
Flash Point 89°C (192°F)
Chemical Name or Material 2,4-Dichloropyridine
Beilstein 108666
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P280h-P305+P351+P338-P361-P309+P311-P302+P352
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
IF exposed or if you feel unwell: Call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.

GHS H Statement
H311-H227-H302-H315-H319-H335
Toxic in contact with skin.
Combustible liquid.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.