Please login to your online account to display your discounted pricing

Alfa Aesar™ 2',4'-Dichloropropiophenone, 97%

$250.43 - $250.43

Chemical Identifiers

Chemical Name or Material 2',4'-Dichloropropiophenone
CAS 37885-41-9
Molecular Formula C9H8Cl2O
Formula Weight 203.07g/mol
MDL Number MFCD00027396
Beilstein 2248270
Synonym 1-(2,4-Dichlorophenyl)-1-propanone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1125222

 
alfa aesar™
A11252-22
100g Each for $250.43
Description & Specifications

Specifications

Boiling Point 121° to 123°C (7mmHg)
Flash Point >110°C (230°F)
Quantity 100g
Chemical Name or Material 2',4'-Dichloropropiophenone
CAS 37885-41-9
Assay 97%
Molecular Formula C9H8Cl2O
Formula Weight 203.07g/mol
MDL Number MFCD00027396
Beilstein 2248270
Synonym 1-(2,4-Dichlorophenyl)-1-propanone
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for sev

Applications
2′,4′-Dichloropropiophenone is used in the synthesis of (S)-3-chloro-1-(4-chlorophenyl)-1-propanol.

Solubility
Slightly soluble in water

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.