Please login to your online account to display your discounted pricing

2,4-Dichlorophenylhydrazine hydrochloride, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2, 4-Dichlorophenylhydrazine hydrochloride
CAS 5446-18-4
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C6H6Cl2N2·ClH
Linear Formula Cl2C6H3NHNH2·HCl
Formula Weight 213.49
MDL Number MFCD00012929
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC166740250

  SDS
thermo fisher scientific
166740250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Formula Weight 213.49
Physical Form Crystalline Powder
Quantity 25g
Color Off-white to light brown
Beilstein 15, I, 107
CAS Max % 100.0
Linear Formula Cl2C6H3NHNH2·HCl
Infrared Spectrum Authentic
CAS 5446-18-4
Molecular Formula C6H6Cl2N2·ClH
Health Hazard 3 GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. Do not breathe dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
CAS Min % 98.5
Packaging Glass bottle
Chemical Name or Material 2, 4-Dichlorophenylhydrazine hydrochloride
MDL Number MFCD00012929
Assay Percent Range 99%
Health Hazard 1 Warning
Health Hazard 2 GHS P Statement Toxic if swallowed. Toxic in contact with skin. Toxic if inhaled. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.