missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,4-Dichlorophenylhydrazine hydrochloride, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 166740250

 View more versions of this product

Catalog No. AC166740250

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

2, 4-Dichlorophenylhydrazine hydrochloride
25g
Authentic
100.0
Crystalline Powder
99%
Cl2C6H3NHNH2·HCl
MFCD00012929
Off-white to light brown
5446-18-4
98.5
Glass bottle
≥98.5%
C6H6Cl2N2·ClH
213.49
15, I, 107

Chemical Identifiers

5446-18-4
100.0
C6H6Cl2N2·ClH
213.49
2, 4-Dichlorophenylhydrazine hydrochloride
98.5
99%
Cl2C6H3NHNH2·HCl
MFCD00012929
≥98.5%

Safety and Handling

SDS & Safety Documents

GHS P Statement Toxic if swallowed. Toxic in contact with skin. Toxic if inhaled. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. Do not breathe dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning