missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,4-Dichlorophenylboronic acid, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC367670010
Specifications
2, 4-Dichlorophenylboronic acid | |
Glass bottle | |
240.0°C to 245.0°C | |
97% | |
96.0 | |
97% | |
MFCD00013930 | |
Solubility in water: insoluble. Other solubilities: soluble in methanol | |
B(C1=C(C=C(C=C1)Cl)Cl)(O)O | |
190.82 | |
190.82 | |
97% |
Authentic | |
White | |
1g | |
68716-47-2 | |
100.0 | |
C6H5BCl2O2 | |
2,4-dichloro benzene boronic acid, 2,4-dichlorobenzeneboron dihydroxide, 2,4-dichlorobenzeneboronic acid, 2,4-dichlorobenzeneboronicacid, 2,4-dichlorophenyl boranediol, 2,4-dichlorophenyl boronic acid, 2,4-dichlorophenylbornic acid, boronic acid, 2,4-dichlorophenyl, chembl20726 | |
QNEGDGPAXKYZHZ-UHFFFAOYSA-N | |
(2,4-dichlorophenyl)boronic acid | |
2734659 | |
Powder |
Chemical Identifiers
68716-47-2 | |
190.82 | |
QNEGDGPAXKYZHZ-UHFFFAOYSA-N | |
2734659 | |
B(C1=C(C=C(C=C1)Cl)Cl)(O)O |
C6H5BCl2O2 | |
MFCD00013930 | |
2,4-dichloro benzene boronic acid, 2,4-dichlorobenzeneboron dihydroxide, 2,4-dichlorobenzeneboronic acid, 2,4-dichlorobenzeneboronicacid, 2,4-dichlorophenyl boranediol, 2,4-dichlorophenyl boronic acid, 2,4-dichlorophenylbornic acid, boronic acid, 2,4-dichlorophenyl, chembl20726 | |
(2,4-dichlorophenyl)boronic acid |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face prot
GHS Signal Word: Warning
Recommended Storage : Refrigerator +4°C