Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Dichlorophenylacetonitrile, 98+%

Chemical Identifiers

Chemical Name or Material 2,4-Dichlorophenylacetonitrile
CAS 6306-60-1
Molecular Formula C8H5Cl2N
Formula Weight 186.04
MDL Number MFCD00001899
UN Number UN3439
Beilstein 2047688
Synonym 2,4-Dichlorobenzyl cyanide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1287322

  SDS
alfa aesar™
A1287322
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 186.04
Assay ≥98%
Synonym 2,4-Dichlorobenzyl cyanide
Health Hazard 1 Danger
Molecular Formula C8H5Cl2N
MDL Number MFCD00001899
UN Number UN3439
Chemical Name or Material 2,4-Dichlorophenylacetonitrile
Quantity 100g
Odor Characteristic
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
Beilstein 2047688
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Flash Point 176°C (348°F)
Melting Point 58° to 62°C
Health Hazard 2 GHS H Statement
H301-H311-H331-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS 6306-60-1
Safety and Handling

Danger

GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H301-H311-H331-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.