Please login to your online account to display your discounted pricing

2,4-Dichlorophenylacetone, 98%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  35081-0010

 View more versions of this product

Catalog No. AC350810010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Formula Weight 203.07
Physical Form Liquid After Melting
Quantity 1g
Color Clear light yellow
TSCA TSCA
CAS Max % 100.0
CAS Min % 97.5
Refractive Index 1.5485 to 1.5505 (20°C, 589nm)
Packaging Glass bottle
Infrared Spectrum Authentic
Name Note 98%
CAS 93457-07-9
MDL Number MFCD00210405
Chemical Name or Material 2, 4-Dichlorophenylacetone
Molecular Formula C9H8Cl2O
Assay Percent Range 98%

Specifications

Chemical Name or Material 2, 4-Dichlorophenylacetone
Name Note 98%
CAS 93457-07-9
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C9H8Cl2O
Formula Weight 203.07
MDL Number MFCD00210405