Please login to your online account to display your discounted pricing

2,4-Dichlorophenylacetone, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2, 4-Dichlorophenylacetone
Name Note 98%
CAS 93457-07-9
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C9H8Cl2O
Formula Weight 203.07
MDL Number MFCD00210405
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC350810010

  SDS
thermo fisher scientific
35081-0010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC350810050

  SDS
thermo fisher scientific
35081-0050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Liquid After Melting
Formula Weight 203.07
Quantity 1g
Color Clear light yellow
TSCA TSCA
CAS Max % 100.0
Chemical Name or Material 2, 4-Dichlorophenylacetone
Infrared Spectrum Authentic
Name Note 98%
Assay Percent Range 98%
Molecular Formula C9H8Cl2O
MDL Number MFCD00210405
Packaging Glass bottle
CAS 93457-07-9
Refractive Index 1.5485 to 1.5505 (20°C, 589nm)
CAS Min % 97.5