Please login to your online account to display your discounted pricing

2,4-Dichlorophenylacetic Acid 99%, ACROS Organics™

$43.91 - $43.91

Chemical Identifiers

Chemical Name or Material 2, 4-Dichlorophenylacetic acid
Name Note 99%
CAS 19719-28-9
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C8H6Cl2O2
Linear Formula Cl2C6H3CH2COOH
Formula Weight 205.04
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC134270100

  SDS
thermo fisher scientific
134270100
Glass bottle 10g Each for $43.91
Description & Specifications

Specifications

Physical Form Crystalline Powder
Assay ≥98.5%
Formula Weight 205.04
Quantity 10g
Color White to pale yellow-beige
EINECS Number 243-248-7
TSCA TSCA
Melting Point 128° to 133°C
Beilstein 09, III, 2271
CAS Max % 100.0
Chemical Name or Material 2, 4-Dichlorophenylacetic acid
Infrared Spectrum Authentic
Name Note 99%
Assay Percent Range 99%
Molecular Formula C8H6Cl2O2
Linear Formula Cl2C6H3CH2COOH
MDL Number MFCD00004318
Packaging Glass bottle
CAS 19719-28-9
CAS Min % 98.5
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.