Please login to your online account to display your discounted pricing

2,4-Dichlorophenyl isothiocyanate, 95%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2, 4-Dichlorophenyl isothiocyanate
Name Note 95%
CAS 6590-96-1
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C7H3Cl2NS
Linear Formula Cl2C6H3NCS
Formula Weight 204.08
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC305280050

  SDS
thermo fisher scientific
305280050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥94%
Physical Form Crystalline Solid
Formula Weight 204.08
Quantity 5g
Color White
Density 1.4100g/mL
EINECS Number 229-527-6
TSCA TSCA
Melting Point 36° to 42°C
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Molecular Formula C7H3Cl2NS
Linear Formula Cl2C6H3NCS
CAS 6590-96-1
Infrared Spectrum Authentic
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Harmful if inhaled.
Harmful if swallowed.
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 2, 4-Dichlorophenyl isothiocyanate
Name Note 95%
MDL Number MFCD00037827
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Harmful if inhaled.
Harmful if swallowed.
Causes severe skin burns and eye damage.
Harmful in contact with skin.