Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Dichlorophenoxyacetic acid hydrazide, 97%

$60.96 - $184.54

Chemical Identifiers

Chemical Name or Material 2,4-Dichlorophenoxyacetic acid hydrazide
CAS 28236-62-6
Molecular Formula C8H8Cl2N2O2
Formula Weight 235.07g/mol
MDL Number MFCD00051972
Beilstein 986266
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1551306

 
alfa aesar™
A15513-06
5g Each for $60.96

AAA1551314

 
alfa aesar™
A15513-14
25g Each for $184.54
Description & Specifications

Specifications

Melting Point 150° to 152°C
Quantity 5g
Chemical Name or Material 2,4-Dichlorophenoxyacetic acid hydrazide
CAS 28236-62-6
Assay 97%
Molecular Formula C8H8Cl2N2O2
Formula Weight 235.07g/mol
MDL Number MFCD00051972
Beilstein 986266
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.