Learn More
2,4-Dichlorophenethylamine, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 350780050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
2, 4-Dichlorophenethylamine | |
52516-13-9 | |
100.0 | |
>110°C | |
97.5% min. (GC) | |
C8H10Cl2N | |
Cl2C6H3CH2CH2NH2 | |
5g | |
1.26 | |
VHJKDOLGYMULOP-UHFFFAOYSA-O | |
2-(2,4-dichlorophenyl)ethan-1-aminium | |
142938 | |
98% |
98% | |
97.5 | |
1.2600g/mL | |
Authentic | |
Glass bottle | |
1.5645 to 1.5665 | |
MFCD00060611 | |
12,IV,2491 | |
2,4-dichlorophenethylamine, 2-2,4-dichlorophenyl ethanamine, 2-2,4-dichlorophenyl ethylamine, 2-2,4-dichlorophenyl ethan-1-amine, benzeneethanamine, 2,4-dichloro, acmc-209kzz, 2,4-dichlorophenethyl amine, ksc497e8j, benzeneethanamine,2,4-dichloro | |
[NH3+]CCC1=CC=C(Cl)C=C1Cl | |
191.07 | |
190.07 |
Chemical Identifiers
52516-13-9 | |
191.07 | |
VHJKDOLGYMULOP-UHFFFAOYSA-O | |
142938 | |
[NH3+]CCC1=CC=C(Cl)C=C1Cl |
C8H10Cl2N | |
MFCD00060611 | |
2,4-dichlorophenethylamine, 2-2,4-dichlorophenyl ethanamine, 2-2,4-dichlorophenyl ethylamine, 2-2,4-dichlorophenyl ethan-1-amine, benzeneethanamine, 2,4-dichloro, acmc-209kzz, 2,4-dichlorophenethyl amine, ksc497e8j, benzeneethanamine,2,4-dichloro | |
2-(2,4-dichlorophenyl)ethan-1-aminium |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present
GHS Signal Word: Warning
TSCA : TSCA
RUO â Research Use Only