Please login to your online account to display your discounted pricing

2,4-Dichlorophenethylamine, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2, 4-Dichlorophenethylamine
Name Note 98%
CAS 52516-13-9
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C8H9Cl2N
Linear Formula ClC6H3CHCHNH
Formula Weight 190.07
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC350780250

  SDS
thermo fisher scientific
35078-0250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC350780050

  SDS
thermo fisher scientific
35078-0050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC350781000

  SDS
thermo fisher scientific
AC350781000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Liquid
Formula Weight 190.07
Quantity 25g
Color Clear colorless to yellow
TSCA TSCA
Density 1.26g/mL
CAS Max % 100.0
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
CAS Min % 97.5
Refractive Index 1.5645 to 1.5665 (20°C, 589nm)
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Molecular Formula C8H9Cl2N
Health Hazard 1 GHS Signal Word: Danger
CAS 52516-13-9
MDL Number MFCD00060611
Assay Percent Range 98%
Name Note 98%
Chemical Name or Material 2, 4-Dichlorophenethylamine
Linear Formula ClC6H3CHCHNH
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.