missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,4-Dichloronitrobenzene 97%, ACROS Organics™

Manufacturer:  Acros Organics 113520010

 View more versions of this product

Catalog No. AC113520010



Specifications

Specifications

2, 4-Dichloronitrobenzene
Glass bottle
28°C to 33°C
97%
96.0
97%
Cl2C6H3NO2
05, 245
QUIMTLZDMCNYGY-UHFFFAOYSA-N
2,4-dichloro-1-nitrobenzene
11899
Crystalline Low Melting Solid
Authentic
Brown-Yellow
1kg
611-06-3
100.0
C6H3Cl2NO2
MFCD00007071
2,4-dichloronitrobenzene, 1,3-dichloro-4-nitrobenzene, benzene, 2,4-dichloro-1-nitro, unii-85p1a896rr, ccris 3095, benzene, 1,3-dichloro-4-nitro, nitro-m-dichlorobenzene, pubchem15890, 2,4-dichloronitrobenzen, 4-nitro-m-dichlorobenzene
C1=CC(=C(C=C1Cl)Cl)[N+](=O)[O-]
191.995
192
≥96.0%

Chemical Identifiers

611-06-3
191.995
QUIMTLZDMCNYGY-UHFFFAOYSA-N
11899
C1=CC(=C(C=C1Cl)Cl)[N+](=O)[O-]
C6H3Cl2NO2
MFCD00007071
2,4-dichloronitrobenzene, 1,3-dichloro-4-nitrobenzene, benzene, 2,4-dichloro-1-nitro, unii-85p1a896rr, ccris 3095, benzene, 1,3-dichloro-4-nitro, nitro-m-dichlorobenzene, pubchem15890, 2,4-dichloronitrobenzen, 4-nitro-m-dichlorobenzene
2,4-dichloro-1-nitrobenzene
Safety and Handling

Safety and Handling

GHS H Statement:
Toxic to aquatic life with long lasting effects.
Harmful in contact with skin.
May cause an allergic skin reaction.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Wear eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 210-248-3

RTECSNumber : CZ5420000

TSCA : TSCA

SDS Documents