Please login to your online account to display your discounted pricing

2,4-Dichloronitrobenzene 97%, ACROS Organics™

$73.89 - $221.39

Chemical Identifiers

Chemical Name or Material 2, 4-Dichloronitrobenzene
Name Note 97%
CAS 611-06-3
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C6H3Cl2NO2
Linear Formula Cl2C6H3NO2
Formula Weight 192
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC113520010

  SDS
thermo fisher scientific
113520010
Glass bottle 1kg Each for $221.39

AC113522500

  SDS
thermo fisher scientific
113522500
Glass bottle 250g Each for $73.89
Description & Specifications

Specifications

Physical Form Crystalline Low Melting Solid
Assay ≥96.0%
Formula Weight 192
Quantity 1kg
Color Yellow-brown
Beilstein 05, 245
EINECS Number 210-248-3
TSCA TSCA
RTECS Number CZ5420000
Melting Point 28° to 33°C
CAS Min % 96.0
CAS Max % 100.0
Packaging Glass bottle
Health Hazard 1 GHS Signal Word: Warning
Infrared Spectrum Authentic
Name Note 97%
Synonym 1, 3-Dichloro-4-nitrobenzene
Health Hazard 2 GHS H Statement:
Toxic to aquatic life with long lasting effects.
Harmful in contact with skin.
May cause an allergic skin reaction.
Harmful if swallowed.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Wear eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 2, 4-Dichloronitrobenzene
Assay Percent Range 97%
Molecular Formula C6H3Cl2NO2
Linear Formula Cl2C6H3NO2
MDL Number MFCD00007071
CAS 611-06-3
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Wear eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic to aquatic life with long lasting effects.
Harmful in contact with skin.
May cause an allergic skin reaction.
Harmful if swallowed.