Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Dichlorobenzyl mercaptan, 98+%

Chemical Identifiers

Chemical Name or Material 2,4-Dichlorobenzyl mercaptan
CAS 59293-67-3
Molecular Formula C7H6Cl2S
Formula Weight 193.1
MDL Number MFCD00039650
UN Number UN2810
Beilstein 509602
Synonym 2,4-Dichloro-alpha-toluenethiol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2499906

  SDS
alfa aesar™
B2499906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2499914

  SDS
alfa aesar™
B2499914
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 193.1
UN Number UN2810
MDL Number MFCD00039650
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
CAS 59293-67-3
Synonym 2,4-Dichloro-alpha-toluenethiol
Boiling Point 150° to 151°C (12mmHg)
Health Hazard 1 Danger
Chemical Name or Material 2,4-Dichlorobenzyl mercaptan
Assay ≥98%
Flash Point >100°C (212°F)
Molecular Formula C7H6Cl2S
Beilstein 509602
Health Hazard 3 GHS P Statement
P260-P280h-P305+P351+P338-P309-P310
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Health Hazard 2 GHS H Statement
H331-H302-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 5g
Safety and Handling

Danger

GHS P Statement
P260-P280h-P305+P351+P338-P309-P310
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H331-H302-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.