missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,4-Dichlorobenzyl isocyanate, 98%, ACROS Organics™

Chemical Identifiers

CAS 19654-32-1
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C8H5Cl2NO
Formula Weight 202.04
Chemical Name or Material 2, 4-Dichlorobenzyl isocyanate
Assay ≥97.5% (GC)
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC440350100 SDS Thermo Fisher Scientific
AC440350100
10g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC440350020 SDS Thermo Fisher Scientific
AC440350020
2g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

19654-32-1
100.0
C8H5Cl2NO
2, 4-Dichlorobenzyl isocyanate
97.5
98%
202.04
≥97.5% (GC)

Specifications

Crystalline Powder
10g
1.388g/mL
19654-32-1
97.5
Authentic
≥97.5% (GC)
202.04
White to yellow to light brown
100.0
C8H5Cl2NO
98%
2, 4-Dichlorobenzyl isocyanate

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning