Learn More
2,4-Dichlorobenzoyl chloride, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 150220050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
2, 4-Dichlorobenzoyl chloride | |
89-75-8 | |
100.0 | |
150°C (34.0mmHg) | |
Authentic | |
Glass bottle | |
1.5915 to 1.5937 | |
MFCD00000661 | |
09, 342 | |
benzoyl chloride, 2,4-dichloro, 2,4-dichlorobenzoylchloride, benzoylchloride, 2,4-dichloro, ccris 8616, acmc-1cuf7, dsstox_cid_31176, dsstox_gsid_52603, 2,4-dichloro benzoylchloride, 2.4-dichlorobenzoyl chloride, ksc450c2p | |
CEOCVKWBUWKBKA-UHFFFAOYSA-N | |
2,4-dichlorobenzoyl chloride | |
66645 | |
98% |
98% | |
97.5 | |
1.4900g/mL | |
137°C | |
97.5% min. (GC) | |
C7H3Cl3O | |
Cl2C6H3COCl | |
5mL | |
1.49 | |
Solubility in water: may decompose. Other solubilities: soluble in alcohol,ether and acetone,slightly soluble in heptane | |
ClC(=O)C1=CC=C(Cl)C=C1Cl | |
209.45 | |
209.46 |
Chemical Identifiers
89-75-8 | |
209.45 | |
CEOCVKWBUWKBKA-UHFFFAOYSA-N | |
66645 | |
ClC(=O)C1=CC=C(Cl)C=C1Cl |
C7H3Cl3O | |
MFCD00000661 | |
benzoyl chloride, 2,4-dichloro, 2,4-dichlorobenzoylchloride, benzoylchloride, 2,4-dichloro, ccris 8616, acmc-1cuf7, dsstox_cid_31176, dsstox_gsid_52603, 2,4-dichloro benzoylchloride, 2.4-dichlorobenzoyl chloride, ksc450c2p | |
2,4-dichlorobenzoyl chloride |
Safety and Handling
GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 201-936-4
RTECSNumber : DM6636733
TSCA : TSCA
RUO â Research Use Only